Please login to your online account to display your discounted pricing

anti-DAAM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224536SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224536SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 23500
Primary or Secondary Primary
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human and rhesus monkey, 94% homologous in horse, 88% homologous in dog, and 76% homologous in cow, mouse and rat. For IF, the GLI1 IMG-6553A antibody was used at 10 ug/ml (Hu, 2010). Protocol: Cells were washed in PBS and fixed in 4% PFA, pH 7.4 (20g PFA in 500 ml PBS) for 10 min. The fixed cells were permeabilized in 0.5% Triton X-100 (in PBS) for 5 min and blocking in 50 ul of 20% HINGS (heat inactivated normal goat serum diluted in PBS) for 1 hr. The GLI1 antibody was incubated at 37 degrees C for 1 hr or at 4 degrees C overnight. Following 3 X 5 min washes in PBS, an AlexaFluor488 or 594-conjugated goat anti-rabbit antibody was used as a second step (diluted in 20% HINGS at 1:1000 for 1 hr at room temperature i the dark). Following 3 X 5 min washes in PBS, the slides were mounted with DABCO (1,4-diazabicyclo[2.2.2]octane) anti-fading agent for fluorescence microscopy.
Concentration 0.5mg/mL
Host Species Rabbit
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen DAAM2
Immunogen A portion of amino acids 100-150 of human DAAM2 was used as the immunogen for this antibody.
Quantity 0.025mg
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Affinity Purified