Please login to your online account to display your discounted pricing

anti-DAAM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224623SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224623SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 23002
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 2) and 87% homologous in mouse.
Gene Accession No. Q9Y4D1
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen DAAM1
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Quantity 0.025mg
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Immunogen A portion of amino acids 1-50 of human DAAM1 was used as the immunogen.
Host Species Rabbit
Format Purified