Please login to your online account to display your discounted pricing

anti-DAAM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224623

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224623

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 23002
Gene Accession No. Q9Y4D1
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 2) and 87% homologous in mouse.
Regulatory Status RUO
Format Purified
Immunogen A portion of amino acids 1-50 of human DAAM1 was used as the immunogen.
Host Species Rabbit
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen DAAM1
Quantity 0.1mg