Please login to your online account to display your discounted pricing

anti-DAAM1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human DAAM1 was used as the immunogen.
Antigen DAAM1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224623SS

 
novus biologicals canada
NBP224623SS
Unlabeled 0.025mg Each for $109.00

NBP224623

 
novus biologicals canada
NBP224623
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human DAAM1 was used as the immunogen.
Antigen DAAM1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 2) and 87% homologous in mouse.
Gene Accession No. Q9Y4D1
Gene ID (Entrez) 23002
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL