missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Cytokeratin 19, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Cytokeratin 19 protein (between residues 1-50) [UniProt P08727]
Antigen Cytokeratin 19
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178278SS Novus Biologicals Canada
NBP178278SS
0.025mL Each for $139.00
NBP178278 Novus Biologicals Canada
NBP178278
0.1mL Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
Cytokeratin 19
Affinity Purified
RUO
P08727
PBS, 30% glycerol
A synthetic peptide made to an N-terminal portion of the human Cytokeratin 19 protein (between residues 1-50) [UniProt P08727]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Simple Western,Western Blot
3880
1.16mg/mL