Please login to your online account to display your discounted pricing

anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™

$111.78 - $307.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human CYR61. [UniProt No. O00622]
Antigen Cyr61/CCN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100358

 
novus biologicals canada
NB100358
Unlabeled 0.1mL Each for $307.00

NB100358SS

 
novus biologicals canada
NB100358SS
Unlabeled 0.025mL Each for $111.78
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human CYR61. [UniProt No. O00622]
Antigen Cyr61/CCN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human. Immunogen sequence has 99% homology to Xenopus, 93% homology to chicken and 80% homology to rat and mouse.
Gene Accession No. O00622
Gene ID (Entrez) 3491
Formulation 50% Glycerol, 1% BSA, PBS, pH 7.2
Concentration 1mg/mL