Please login to your online account to display your discounted pricing

anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™

$160.00 - $332.66

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 250-350). [UniProt No. O00622]
Antigen Cyr61/CCN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100357

 
novus biologicals canada
NB100357
Unlabeled 0.1mL Each for $332.66

NB100357SS

 
novus biologicals canada
NB100357SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 250-350). [UniProt No. O00622]
Antigen Cyr61/CCN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity Human. Immunogen sequence has 100% homology to rat and 96% homology to mouse protein; however we have had negative feedback with the use of this antibody in mouse and rat tissue.
Gene Accession No. O00622
Gene ID (Entrez) 3491
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL