Please login to your online account to display your discounted pricing

anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). [UniProt No. O00622]
Antigen Cyr61/CCN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100356SS

 
novus biologicals canada
NB100356SS
Unlabeled 0.025mL Each for $144.63

NB100356

 
novus biologicals canada
NB100356
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). [UniProt No. O00622]
Antigen Cyr61/CCN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Neutralization
Applications Simple Western
Applications Western Blot
Test Specificity Human. The immunogen sequence has 88% homology to rat and mouse.
Gene Accession No. O00622
Gene ID (Entrez) 3491
Formulation PBS
Concentration 1mg/mL