Please login to your online account to display your discounted pricing

anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100358SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100358SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Formulation 50% Glycerol, 1% BSA, PBS, pH 7.2
Gene Accession No. O00622
Primary or Secondary Primary
Gene ID (Entrez) 3491
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Test Specificity Human. Immunogen sequence has 99% homology to Xenopus, 93% homology to chicken and 80% homology to rat and mouse.
Host Species Rabbit
Quantity 0.025mL
Cross Reactivity Human
Immunogen A synthetic peptide made to the C-terminal region of human CYR61. [UniProt No. O00622]
Monoclonal or Polyclonal Polyclonal
Antigen Cyr61/CCN1
Format Affinity Purified