Please login to your online account to display your discounted pricing

anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100358

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100358

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. O00622
Formulation 50% Glycerol, 1% BSA, PBS, pH 7.2
Test Specificity Human. Immunogen sequence has 99% homology to Xenopus, 93% homology to chicken and 80% homology to rat and mouse.
Format Affinity Purified
Gene ID (Entrez) 3491
Concentration 1mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Regulatory Status RUO
Immunogen A synthetic peptide made to the C-terminal region of human CYR61. [UniProt No. O00622]
Cross Reactivity Human
Antigen Cyr61/CCN1
Host Species Rabbit