Please login to your online account to display your discounted pricing

anti-Cyr61/CCN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100357SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100357SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3491
Gene Accession No. O00622
Formulation Tris-citrate/phosphate, pH 7-8
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Format Affinity Purified
Concentration 1mg/mL
Test Specificity Human. Immunogen sequence has 100% homology to rat and 96% homology to mouse protein; however we have had negative feedback with the use of this antibody in mouse and rat tissue.
Antigen Cyr61/CCN1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Host Species Rabbit
Primary or Secondary Primary
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 250-350). [UniProt No. O00622]