Please login to your online account to display your discounted pricing

anti-CYLD, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11095574SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11095574T

  • / Each
 

Description & Specifications

Specifications

Antigen CYLD
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q9NQC7
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human CYLD (within residues 500-600). [Swiss-Prot No. Q9NQC7]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1540
Test Specificity Human, bovine, orangutan and mouse. Does not react with rat.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Primate