Learn More
cytochrome c, somatic, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CYCS protein.
Supplier: Abnova Corporation H00054205B01
Description
This gene encodes cytochrome c, a component of the electron transport chain in mitochondria. The heme group of cytochrome c accepts electrons from the b-c1 complex and transfers electrons to the cytochrome oxidase complex. Cytochrome c is also involved in initiation of apoptosis. Upon release of cytochrome c to the cytoplasm, the protein binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Many cytochrome c pseudogenes exist, scattered throughout the human genome. [provided by RefSeq
Sequence: MGDVEKGKKIFIMKCSQCHTVEKGGKHKTGPNLHGLFGRKTGQAPGYSYTAANKNKGIIWGEDTLMEYLENPKKYIPGTKMIFVGIKKKEERADLIAYLKKATNESpecifications
cytochrome c, somatic | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human CYCS protein. | |
CYCS | |
CYC/HCS | |
Mouse | |
50 μL | |
Yes | |
54205 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC005299 | |
CYCS | |
CYCS (AAH05299, 1 a.a. ∼ 105 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |