Please login to your online account to display your discounted pricing

anti-Cyclophilin B, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The portions of amino acids range between 100-228 of human Cyclophilin B was used as the immunogen.
Antigen Cyclophilin B
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224770SS

 
novus biologicals canada
NBP224770SS
Unlabeled 0.025mg Each for $109.00

NBP224770

 
novus biologicals canada
NBP224770
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The portions of amino acids range between 100-228 of human Cyclophilin B was used as the immunogen.
Antigen Cyclophilin B
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog and bovine, and 84% in chicken.
Gene Accession No. P23284
Gene ID (Entrez) 5479
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL