Please login to your online account to display your discounted pricing

anti-Cyclin D1, Polyclonal, Novus Biologicals™

$160.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 250 to 300 of human Cyclin D1 was used as immunogen.
Antigen Cyclin D1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224695

 
novus biologicals canada
NBP224695
Unlabeled 0.1mg Each for $307.00

NBP224695SS

 
novus biologicals canada
NBP224695SS
Unlabeled 0.025mg Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 250 to 300 of human Cyclin D1 was used as immunogen.
Antigen Cyclin D1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The eIF3J amino acid sequence used as immunogen for this antibody is 100% homologous in human, horse, chimpanzee, rat, dog, mouse, orangutan, opossum and platypus, 94% homologous in cow, zebrafish and chicken and 76% homologous in Tetradon.
Gene ID (Entrez) 595
Formulation 100μg of antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL