Please login to your online account to display your discounted pricing

anti-cyclic GMP, Polyclonal, Alfa Aesar™

Rabbit Polyclonal Antibody

Specifications

Antigen cyclic GMP
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Host Species Rabbit
Format Lyophilized
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64025AMO

 
alfa aesar
J64025AMO
1000 Tests CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ64025KX

 
alfa aesar
J64025KX
100 Tests CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Antigen cyclic GMP
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Host Species Rabbit
Format Lyophilized
Conjugate Unlabeled
Storage Requirements 2-8°C
Applications RadioImmunoassay