Please login to your online account to display your discounted pricing

anti-CXXC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056630

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056630

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9P0U4
Gene ID (Entrez) 30827
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Concentration 0.5mg/mL
Cross Reactivity Human
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Regulatory Status RUO
Applications Western Blotting
Format Purified
Quantity 0.2mL
Host Species Rabbit
Antigen CXXC1
Immunogen This polyclonal antibody was raised against a mixture of synthetic peptides containing 73-90 and 641-656 amino acids residues of human CGBP.