missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CXXC1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a mixture of synthetic peptides containing 73-90 and 641-656 amino acids residues of human CGBP.
Antigen CXXC1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056630 Novus Biologicals Canada
NB10056630
0.2mL Each for $255.80
NB10056630T Novus Biologicals Canada
NB10056630SS
0.05mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
CXXC1
Purified
RUO
Q9P0U4
PBS containing 0.05% BSA
This polyclonal antibody was raised against a mixture of synthetic peptides containing 73-90 and 641-656 amino acids residues of human CGBP.
Polyclonal
Primary
Western Blot
30827
0.5mg/mL