Please login to your online account to display your discounted pricing

anti-CXXC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056630SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056630T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Quantity 0.05mL
Gene ID (Entrez) 30827
Concentration 0.5mg/mL
Gene Accession No. Q9P0U4
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Immunogen This polyclonal antibody was raised against a mixture of synthetic peptides containing 73-90 and 641-656 amino acids residues of human CGBP.
Antigen CXXC1