Please login to your online account to display your discounted pricing

anti-CXCR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056333SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056333T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_116743.1
Gene ID (Entrez) 643
Conjugate Unlabeled
Concentration 0.5mg/mL
Formulation PBS
Regulatory Status RUO
Test Specificity A 36 kDa band should be observed.
Host Species Rabbit
Cross Reactivity Human
Antigen CXCR5
Quantity 0.025mg
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Immunogen This antibody was raised against a synthetic peptide of human CXCR5 protein corresponding to amino acids 20-32.
Applications Western Blotting
Format Purified