Please login to your online account to display your discounted pricing

anti-CXCR5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056333

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056333

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene ID (Entrez) 643
Gene Accession No. NP_116743.1
Test Specificity A 36 kDa band should be observed.
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Format Purified
Immunogen This antibody was raised against a synthetic peptide of human CXCR5 protein corresponding to amino acids 20-32.
Applications Western Blotting
Conjugate Unlabeled
Quantity 0.1mg
Cross Reactivity Human
Regulatory Status RUO
Primary or Secondary Primary
Host Species Rabbit
Antigen CXCR5