Please login to your online account to display your discounted pricing

anti-CXCR5, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was raised against a synthetic peptide of human CXCR5 protein corresponding to amino acids 20-32.
Antigen CXCR5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056333

 
novus biologicals canada
NB10056333
Unlabeled 0.1mg Each for $255.80

NB10056333T

 
novus biologicals canada
NB10056333SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was raised against a synthetic peptide of human CXCR5 protein corresponding to amino acids 20-32.
Antigen CXCR5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity A 36 kDa band should be observed.
Gene Accession No. NP_116743.1
Gene ID (Entrez) 643
Formulation PBS
Concentration 0.5mg/mL