Please login to your online account to display your discounted pricing

anti-CXCR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10074396SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10074396T

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Antiserum
Gene ID (Entrez) 7852
Antigen CXCR4
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.025mL
Immunogen A synthetic peptide made to a C-terminal region of the human CXCR4 protein (within residues 300-352). [Swiss-Prot P61073]
Regulatory Status RUO
Primary or Secondary Primary
Host Species Rabbit