Please login to your online account to display your discounted pricing

anti-CXCR4, Polyclonal, Novus Biologicals™

$109.00 - $362.00

Specifications

Host Species Rabbit
Immunogen Rabbit anti-CXCR4 polyclonal antibody was raised against a peptide corresponding to amino acids 328-338 of human CXCR4.
Antigen CXCR4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056437

 
novus biologicals canada
NB10056437
Unlabeled 0.1mg Each for $362.00

NB10056437T

 
novus biologicals canada
NB10056437SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Rabbit anti-CXCR4 polyclonal antibody was raised against a peptide corresponding to amino acids 328-338 of human CXCR4.
Antigen CXCR4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In HeLa, a 45 kDa band is observed.
Gene Accession No. P61073
Gene ID (Entrez) 7852
Formulation PBS with 0.05% BSA
Concentration 0.5mg/mL