Please login to your online account to display your discounted pricing

anti-CXCR4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177560

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177560

  • / Each
 

Description & Specifications

Specifications

Applications Simple Western
Applications Western Blotting
Gene Accession No. P61073
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 7852
Immunogen A synthetic peptide made to a C-terminal portion of the human CXCR4 protein (between residues 300-352) [UniProt P61073]
Cross Reactivity Human
Regulatory Status RUO
Quantity 0.1mL
Format Affinity Purified
Primary or Secondary Primary
Host Species Rabbit
Antigen CXCR4