Please login to your online account to display your discounted pricing

anti-CXCR4/CD184 Clone: 12G5, Thermo Scientific™ Novex™ DFS Item


CXCR4/CD184 Monoclonal antibody AHR1462X from Thermo Scientific™ Novex™ specifically detects CXCR4/CD184 in Human and Non-human primate samples, and it is validated for IHC, IHC (F) and FACS

Manufacturer: life technologies  LSAHR1462X

 View more versions of this product

Catalog No. AHR1462X

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Non-Human Primate
Antigen CXCR4/CD184
Format Purified
Formulation 500μg of Protein A/G antibody in PBS with no preservative; pH 7.3.
Gene Accession No. P61073
Host Species Mouse
Immunogen SIV-infected SupT1 cells injected into Balb/c mice.
Isotype IgG2a, κ
Purification Method Protein A/G
Storage Requirements Store at -20°C
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7852
Clone 12G5
Conjugate Unlabeled

Human immunodeficiency virus (HIV) and related viruses require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CXCR4 (fusin, LESTR or HUMSTR) is a principal coreceptor for T-cell tropic strains of HIV-1 fusion and entry of human white blood cells. CXCR4 is also required for the infection by dual-tropic strains of HIV-1 and mediates CD-4 independent infection by HIV-2. The a-chemokine SDF-1 is the ligand for CXCR4 and prevents infection by T-tropic HIV-1. CXCR4 associates with the surface CD4-gp120 complex before HIV enters target cells. CXCR4 messenger RNA levels correlated with HIV-1 permissiveness in diverse human cell types. Antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV biding sites.
Host Species: Mouse
Cross Reactivity: Human, Non-human primate
Isotype: IgG2a, κ
Clone: 12G5
Immunogen: SIV-infected SupT1 cells injected into Balb/c mice.
Formulation: 500μg purified antibody in PBS and no preservative; pH 7.3
Synonyms: D2S201E, FB22, HM89, HSY3RR, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, C-X-C chemokine receptor type 4
Uniprot Accession: P61073
Entrez GeneID: 7852
Gene Symbol: CXCR4
This product detects CXCR4/CD184 in Human and Non-human primate samples. This product has been successfully used in Flow Cytometry, Immunohistochemistry and Immunohistochemistry (Frozen) procedures. The immunogen is SIV-infected SupT1 cells injected into Balb/c mice..

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.