Please login to your online account to display your discounted pricing

anti-CXCR2/IL-8 RB, PerCP, Clone: 48311, R&D Systems

$415.83 - $415.83

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. AAB25880
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

FAB331C100

 
r & d systems
FAB331C100
100 Tests Each for $415.83
Description & Specifications

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. AAB25880

Host Species: Mouse
Clone: 48311
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: NS0 mouse myeloma cell line transfected with human CXCR2/IL-8 RB. Met1-Leu355

Applications:

Flow Cytometry