Please login to your online account to display your discounted pricing

anti-CXCR2/IL-8 RB, PerCP, Clone: 48311, R&D Systems


Manufacturer: r & d systems  FAB331C100

 View more versions of this product

Catalog No. FAB331C100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. AAB25880
Quantity 100 Tests
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate PerCP