missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-CX3CL1/Fractalkine, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human CX3CL protein (within residues 20-150). [Swiss-Prot P78423]
Antigen CX3CL1/Fractalkine
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP149539 Novus Biologicals Canada
NBP149539
0.1mL Each for $329.00
NBP149539SS Novus Biologicals Canada
NBP149539SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
CX3CL1/Fractalkine
Affinity Purified
RUO
P78423
PBS, 30% glycerol
A genomic peptide made to an internal region of the human CX3CL protein (within residues 20-150). [Swiss-Prot P78423]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
6376
0.57mg/mL