Please login to your online account to display your discounted pricing

anti-CTR2, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-Prot No. O15432]
Antigen CTR2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP105199

 
novus biologicals canada
NBP105199
Unlabeled 0.1mL Each for $309.05

NBP105199SS

 
novus biologicals canada
NBP105199SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-Prot No. O15432]
Antigen CTR2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Gene Accession No. O15432
Gene ID (Entrez) 1318
Formulation PBS, 30% glycerol
Concentration 0.93mg/mL