Please login to your online account to display your discounted pricing

anti-CTLA-4, Polyclonal, Novus Biologicals™

$397.00 - $397.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the entire range of the target protein.
Antigen CTLA-4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

1964000201

 
novus biologicals canada
1964000201MG
Unlabeled 0.1mg Each for $397.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the entire range of the target protein.
Antigen CTLA-4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity This product is specific for Human CTLA4.
Gene Accession No. NM_005214
Gene ID (Entrez) 1493
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0