Please login to your online account to display your discounted pricing

anti-CTGF/CCN2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProt No. P29279]
Antigen CTGF/CCN2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100724SS

 
novus biologicals canada
NB100724SS
Unlabeled 0.025mL Each for $139.00

NB100724

 
novus biologicals canada
NB100724
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProt No. P29279]
Antigen CTGF/CCN2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Test Specificity Human, mouse and rat. Predicted to react with porcine, bovine, chicken and sheep based on 100% sequence homology.
Gene Accession No. P29279
Gene ID (Entrez) 1490
Formulation Tris-glycine, 150mM NaCl
Concentration 1.01mg/mL