Please login to your online account to display your discounted pricing

anti-CTGF/CCN2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100724

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100724

  • / Each
 

Description & Specifications

Specifications

Antigen CTGF/CCN2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Concentration 1.01mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P29279
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProt No. P29279]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1490
Test Specificity Human, mouse and rat. Predicted to react with porcine, bovine, chicken and sheep based on 100% sequence homology.
Target Species Human
Target Species Murine
Target Species Rat