Please login to your online account to display your discounted pricing

anti-CTCF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056494

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056494

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10664
Formulation PBS containing 0.2% gelatin
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c, _d), rhesus monkey, dog, mouse and rat. Cross reacts with Dog, Human, Mouse, Rat and Rhesus Monkey.
Gene Accession No. P49711
Host Species Rabbit
Applications Western Blotting
Concentration 0.5mg/mL
Format Purified
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Immunogen A portion of amino acids 650-700 of human CTCF was used as the immunogen.
Quantity 0.1mg
Conjugate Unlabeled
Primary or Secondary Primary
Antigen CTCF
Monoclonal or Polyclonal Polyclonal