Please login to your online account to display your discounted pricing

anti-CTCF, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human CTCF was used as the immunogen.
Antigen CTCF
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056494

 
novus biologicals canada
NB10056494
Unlabeled 0.1mg Each for $255.80

NB10056494T

 
novus biologicals canada
NB10056494SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 650-700 of human CTCF was used as the immunogen.
Antigen CTCF
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c, _d), rhesus monkey, dog, mouse and rat. Cross reacts with Dog, Human, Mouse, Rat and Rhesus Monkey.
Gene Accession No. P49711
Gene ID (Entrez) 10664
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL