Please login to your online account to display your discounted pricing

anti-CRM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056493SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056493T

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene ID (Entrez) 7514
Test Specificity Reacts with Chimpanzee, Cow, Dog and Human. Predicted cross-reactivity: Mouse and Rat.
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Gene Accession No. O14980
Conjugate Unlabeled
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Immunogen Amino acids 390-408 (ASPLLSGSQHFDVPPRRQL) of human Exportin-1 protein were used as the immunogen (NP_003391).
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.025mg
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Antigen CRM1