Please login to your online account to display your discounted pricing

anti-CRM1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 390-408 (ASPLLSGSQHFDVPPRRQL) of human Exportin-1 protein were used as the immunogen (NP_003391).
Antigen CRM1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056493T

 
novus biologicals canada
NB10056493SS
Unlabeled 0.025mg Each for $139.00

NB10056493

 
novus biologicals canada
NB10056493
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 390-408 (ASPLLSGSQHFDVPPRRQL) of human Exportin-1 protein were used as the immunogen (NP_003391).
Antigen CRM1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Reacts with Chimpanzee, Cow, Dog and Human. Predicted cross-reactivity: Mouse and Rat.
Gene Accession No. O14980
Gene ID (Entrez) 7514
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL