Please login to your online account to display your discounted pricing

anti-CRIPTO, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001597

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001597

  • / Each
 

Description & Specifications

Specifications

Antigen CRIPTO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P13385
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Cripto1 (between residues 100-150). [UniProt No. P13385]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6997
Test Specificity Human. Immunogen has 93% homology with mouse, cow, and Zebrafish protein. There is no significant homology with any other species.
Target Species Human