Please login to your online account to display your discounted pricing

anti-CRIPTO, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Cripto1 (between residues 100-150). [UniProt No. P13385]
Antigen CRIPTO
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001597

 
novus biologicals canada
NB1001597
Unlabeled 0.1mL Each for $329.00

NB1001597SS

 
novus biologicals canada
NB1001597SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Cripto1 (between residues 100-150). [UniProt No. P13385]
Antigen CRIPTO
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human. Immunogen has 93% homology with mouse, cow, and Zebrafish protein. There is no significant homology with any other species.
Gene Accession No. P13385
Gene ID (Entrez) 6997
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL