Please login to your online account to display your discounted pricing

anti-CRF Polyclonal, Thermo Scientific™

CRF Polyclonal antibody PA1-28298 from Thermo Scientific™ specifically detects CRF in Human, Mouse and Rat samples, and it is validated for Western Blot

$651.05 - $651.05

Specifications

Antigen CRF
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PA128298

 
pierce biotechnology inc
PA128298
Each for $651.05
Description & Specifications

Specifications

Antigen CRF
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Chicken
Quantity 50μg
Regulatory Status RUO
Type Primary
Formulation 50μg purified antibody (1mg/mL) in PBS and no preservative
Gene Symbols CRH
Immunogen Synthetic peptide, corresponding to amino acids 27-196 of Human Corticotrophin Releasing Factor CRF.
Isotype IgY
Storage Requirements avoiding freeze/thaw cycles.

CRF Polyclonal antibody PA1-28298 from Thermo Scientific™ specifically detects CRF in Human, Mouse and Rat samples, and it is validated for Western Blot

Corticotropin-releasing hormone (CRH) is a 41-amino acid peptide derived from a 191-amino acid preprohormone. CRH is secreted by the paraventricular nucleus (PVN) of the hypothalamus in response to stress. Marked reduction in CRH has been observed in association with Alzheimer disease and autosomal recessive hypothalamic corticotropin dificiency has multiple and potentially fatal metabolic consequences including hypoglycemia and hepatitis. In addition to production in the hypothalamus, CRH is also synthesized in peripheral tissues, such as T lymphocytes and is highly expressed in the placenta. In the placenta CRH is a marker that determines the length of gestation and the timing of parturition and delivery. A rapid increase in circulating levels of CRH occurs at the onset of parturition, suggesting that, in addition to its metabolic functions, CRH may act as a trigger for parturition.
Host Species: Chicken
Cross Reactivity: Human, Mouse, Rat
Isotype: IgY
Immunogen: Synthetic peptide, corresponding to amino acids 27-196 of Human Corticotrophin Releasing Factor CRF.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with no preservative;
Synonyms: CRF
Uniprot Accession: P01143, P06850, Q8CIT0
Gene Symbol: CRH
Entrez GeneID: 12918, 1392, 81648
PA1-28298 detects CRF in Human, Mouse and Rat samples. PA1-28298 has been successfully used in Western Blot procedures. PA1-28298 The immunogen is Synthetic peptide, corresponding to amino acids 27-196 of Human Corticotrophin Releasing Factor CRF..