Please login to your online account to display your discounted pricing

CRF Polyclonal Antibody, Invitrogen™


CRF Antibody, PA1-28298, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Manufacturer: pierce biotechnology inc  PA128298

Catalog No. PA128298

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified antibody (1mg/mL) in PBS and no preservative
Immunogen Synthetic peptide, corresponding to amino acids 27-196 of Human Corticotrophin Releasing Factor CRF.
Format Purified
Gene Accession No. P01143, P06850, Q8CIT0
Gene Alias CRF
Quantity 50μg
Species Reactivity Human, Mouse, Rat
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 12918, 1392, 81648
Regulatory Status RUO
Antigen CRF
Host Species Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Western Blotting
Isotype IgY
Gene Symbols CRH
Conjugate Unlabeled
Type Primary
Classification Polyclonal

CRF Antibody, PA1-28298, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Corticotropin-releasing hormone (CRH) is a 41-amino acid peptide derived from a 191-amino acid preprohormone. CRH is secreted by the paraventricular nucleus (PVN) of the hypothalamus in response to stress. Marked reduction in CRH has been observed in association with Alzheimer disease and autosomal recessive hypothalamic corticotropin dificiency has multiple and potentially fatal metabolic consequences including hypoglycemia and hepatitis. In addition to production in the hypothalamus, CRH is also synthesized in peripheral tissues, such as T lymphocytes and is highly expressed in the placenta. In the placenta CRH is a marker that determines the length of gestation and the timing of parturition and delivery. A rapid increase in circulating levels of CRH occurs at the onset of parturition, suggesting that, in addition to its metabolic functions, CRH may act as a trigger for parturition.
Host Species: Chicken
Cross Reactivity: Human, Mouse, Rat
Isotype: IgY
Immunogen: Synthetic peptide, corresponding to amino acids 27-196 of Human Corticotrophin Releasing Factor CRF.
Formulation: 50μg purified antibody (1mg/mL) in PBS and no preservative
Synonyms: CRF
Uniprot Accession: P01143, P06850, Q8CIT0
Entrez GeneID: 12918, 1392, 81648
Gene Symbol: CRH
PA1-28298 detects CRF from mouse, rat, human samples. PA1-28298 has been successfully used in Western blot applications. Predicted molecular weight of 70kDa. The PA1-28298 immunogen is: Synthetic peptide, corresponding to amino acids 27-196 of Human Corticotrophin Releasing Factor CRF.