Please login to your online account to display your discounted pricing

anti-CREB (p Ser121), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100410

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100410

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 0.3mg/mL
Gene ID (Entrez) 1385
Formulation Tris 50% glycerol
Monoclonal or Polyclonal Polyclonal
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P16220
Conjugate Unlabeled
Host Species Rabbit
Immunogen A phospho-peptide spanning the Ser121 phosphorylation site of human CREB [UniProt No. P16220]
Antigen CREB (p Ser121)
Quantity 0.1mL
Format Affinity Purified
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse