Please login to your online account to display your discounted pricing

anti-CREB (p Ser121), Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A phospho-peptide spanning the Ser121 phosphorylation site of human CREB [UniProt No. P16220]
Antigen CREB (p Ser121)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100410SS

 
novus biologicals canada
NB100410SS
Unlabeled 0.025mL Each for $144.63

NB100410

 
novus biologicals canada
NB100410
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A phospho-peptide spanning the Ser121 phosphorylation site of human CREB [UniProt No. P16220]
Antigen CREB (p Ser121)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. P16220
Gene ID (Entrez) 1385
Formulation Tris 50% glycerol
Concentration 0.3mg/mL