Please login to your online account to display your discounted pricing

anti-Coronin 3, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 407-426 of human CORO1C.
Antigen Coronin 3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227289

 
novus biologicals canada
NBP227289
Unlabeled 0.1mg Each for $255.80

NBP227289SS

 
novus biologicals canada
NBP227289SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 407-426 of human CORO1C.
Antigen Coronin 3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Gene ID (Entrez) 23603
Formulation 100μg in 200μL PBS containing 0.2% gelatin.
Concentration 0.5mg/mL