Please login to your online account to display your discounted pricing

anti-Coronin-1a, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058867SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058867T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P31146
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Conjugate Unlabeled
Gene ID (Entrez) 11151
Format Antiserum
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Immunogen Full-length recombinant human Coronin-1a expressed in and purified from E. coli. [UniProt No. P31146]
Antigen Coronin-1a