Please login to your online account to display your discounted pricing

anti-Coronin-1a, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058867

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058867

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 11151
Conjugate Unlabeled
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Primary or Secondary Primary
Gene Accession No. P31146
Regulatory Status RUO
Format Antiserum
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Rat
Antigen Coronin-1a
Immunogen Full-length recombinant human Coronin-1a expressed in and purified from E. coli. [UniProt No. P31146]