Please login to your online account to display your discounted pricing

anti-Coronin-1a, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human Coronin-1a expressed in and purified from E. coli. [UniProt No. P31146]
Antigen Coronin-1a
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11058867

 
novus biologicals canada
NB11058867
Unlabeled 0.1mL Each for $329.00

NB11058867T

 
novus biologicals canada
NB11058867SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human Coronin-1a expressed in and purified from E. coli. [UniProt No. P31146]
Antigen Coronin-1a
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Gene Accession No. P31146
Gene ID (Entrez) 11151