missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

delta COP Polyclonal Antibody, Invitrogen™

delta COP Antibody, OSC00035W, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer:  Pierce Biotechnology Inc OSC00035W

Catalog No. PIOSC00035W


 

Specifications

Description

delta COP Antibody, OSC00035W, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from human COPD
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Uniprot Accession: P48444, Q5XJY5, Q66H80
Entrez GeneID: 213827, 300674, 372
Gene Symbol: ARCN1
Reconstitute with 100 ul of distilled water.
Description & Specifications

Specifications

delta COP
Unlabeled
100μL unpurified antibody in whole serum and no preservative
Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Rabbit
100μL
Primary
213827, 300674, 372
Immunohistochemistry,Western Blot
Serum
P48444, Q5XJY5, Q66H80
ARCN1
A synthetic peptide from human COPD
avoiding freeze/thaw cycles.
Polyclonal
Human,Murine,Rat
Promotions

Promotions

  • Purchase any three Invitrogen™ Antibodies and receive a fourth antibody at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    06/30/18