Please login to your online account to display your discounted pricing

anti-delta COP Polyclonal, Thermo Scientific™

delta COP Polyclonal Ab OSC00035W from Thermo Scientific™ specifically detects delta COP in Hu, Ms and Rt samples, and it is validated for IHC and WB

$448.05 - $448.05

Specifications

Antigen delta COP
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00035W

 
pierce biotechnology inc
OSC00035W
Unlabeled 100μL Each for $448.05

PIOSC00036G

 
pierce biotechnology inc
OSC00036G
Unlabeled 500μg Each for $448.05
Description & Specifications

Specifications

Antigen delta COP
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from human COPD
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

delta COP Polyclonal Ab OSC00035W from Thermo Scientific™ specifically detects delta COP in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from human COPD
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Uniprot Accession: P48444, Q5XJY5, Q66H80
Gene Symbol: ARCN1
Entrez GeneID: 213827, 300674, 372
OSC00035W detects delta COP in Human, Mouse and Rat samples. OSC00035W has been successfully used in Immunohistochemistry and Western Blot procedures. OSC00035W The immunogen is A synthetic peptide from human COPD.