Please login to your online account to display your discounted pricing

delta COP Polyclonal Antibody, Invitrogen™


delta COP Antibody, OSC00036G, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00036G

Catalog No. PIOSC00036G

  • / Each
 

Description & Specifications

Specifications

Formulation 500μg purified antibody (1mg/mL) in PBS and no preservative
Applications Immunohistochemistry
Applications Western Blot
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. P48444, Q5XJY5, Q66H80
Gene Alias Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Gene Symbols ARCN1
Storage Requirements avoiding freeze/thaw cycles.
Gene ID (Entrez) 213827, 300674, 372
Species Reactivity Human, Mouse, Rat
Antigen delta COP
Quantity 500μg
Host Species Rabbit
Isotype IgG
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Purified
Immunogen A synthetic peptide from the internal region of human COPD conjugated to an immunogenic carrier protein was used as the antigen

delta COP Antibody, OSC00036G, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: A synthetic peptide from the internal region of human COPD conjugated to an immunogenic carrier protein was used as the antigen
Formulation: 500μg purified antibody (1mg/mL) in PBS and no preservative
Synonyms: Archain, ARCN1, Coatomer subunit delta, Delta-coat protein, Delta-COP
Uniprot Accession: P48444, Q5XJY5, Q66H80
Entrez GeneID: 213827, 300674, 372
Gene Symbol: ARCN1
Reconstitute with 500 ul of distilled water.